Learn More
NGF Mouse anti-Bovine, Human, Mouse, Reptile, Clone: AS21, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112347
Catalog No. 01-674-286
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.
Specifications
Specifications
NGF
Monoclonal
Unconjugated
NGF
Liquid
NGF
Recombit protein corresponding to residues surrounding aa 46-62 of human NGF
Protein A/G
RUO
Antibody
Monoclonal
Bovine, Human, Mouse, Reptile
ELISA, Neutralization
AS21
PBS with 0.08% sodium azide; pH 7.4
P13600, P01138, P01139
Nerve Growth Factor
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
18049, 281350, 4803