Learn More
MilliporeSigma™ anti-Nerve Growth Factor, pain, Clone: 27/21, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB5260Z/DEL
Description
ELISA
Specifications
Nerve Growth Factor, pain | |
Unconjugated | |
2.5 S form of mouse NGF. | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
25 μg | |
Neuroscience | |
Bovine, Human, Mouse, Rat | |
IgG1 |