Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NES1 Mouse anti-Human, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611826

Catalog No. BDB611826


Add to cart

Description

Description

Normal epithelial cell-specific 1 (NES1) was identified in a screen of genes whose mRNA is down-regulated during oncogenic transformation of human mammary epithelial cells (MECs). The protein is homologous to multiple serine proteases including members of the trypsin and kallikrein protease families. Similar to other serine proteases, the sequence of NES1 contains three residues (Ser229, Asp137, and His86) that form a catalytic triad and a putative cleavage site (Arg42). The pattern of NES1 expression in normal human tissues includes prostate, testis, ovary, small intestine, colon, and lung. In addition, expression has been reported to be reduced in human breast and prostate cancer cell lines. NES1 is a secreted protein found in the supernantant of growing MECs. Expression of NES1 in a breast cancer cell line has been reported to suppress oncogenic cell behavior, such as anchorage-independent growth and tumor formation in nude mice. In the breast cancer cell line BT-474, NES1 levels reportedly are up-regulated by steroid hormones, such as estrogen, androgen, and progestin. Thus, NES1 is a steroid-hormone regulated, serine protease that is thought to have important tumor suppressor functions.

Host Species: Mouse
Clone: 2
Isotype: IgG1
Species Reactivity [for Features Main]: Human

Immunofluorescence, Western Blotting

Specifications

Specifications

NES1
2
Unconjugated
Murine
50μg
Cell Biology
Monoclonal
Western Blot
250μg/mL
Affinity Purified
IgG1
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit