Learn More
anti-Nestin, Polyclonal, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 30090002
 View more versions of this product
Catalog No. 30-090-002
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
Nestin
Unlabeled
P48681
Rabbit
0.05mg
Primary
10763
Human
ELISA
20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
Affinity Purified
This antibody is specific for the C Terminus Region of the target protein.
RUO
Polyclonal
This product is specific for Human NES.