Learn More
NET34 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061022
Catalog No. 06-102-2MI
Add to cart
Description
Description
Use Anti-NET34 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry to detect NET34 also known as LIV-1 subfamily of Zimmunoprecipitation zinc transporter 4, Solute carrier family 39 member 14.
Specifications
Specifications
NET34
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human, Murine
Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
SLC39A14; LZT-Hs4; NET34; ZIP14; ZIP-14; cig19
KLH-conjugated linear peptide corresponding to human NET 34 at the extracellular domain.
200μg
Primary
NP_001121903