Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Netrin-1 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB1001605
Description
Netrin-1 Polyclonal specifically detects Netrin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Netrin-1 | |
Polyclonal | |
Unconjugated | |
10mM Sodium PBS (0.9% isotonic, w/v, pH 7.2) with 0.02% Sodium Azide | |
netrin 1, netrin-1, NTN1Lnetrin 1, mouse, homolog of | |
Chicken | |
Affinity Purified | |
RUO | |
9423 | |
Human, Mouse, Rat | |
IgY |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000-1:2000, Immunohistochemistry-Paraffin 1:1000-1:2000, Immunohistochemistry-Frozen | |
O95631, O09118 | |
NTN1 | |
Chickens were immunized with a synthetic peptide-keyhole limpet hemocyanin (KLH) conjugate. This synthetic peptide corresponded to the netrin-1 gene product, but was shared between the human (O_095631, NCBI) and mouse (O_09118, NCBI) sequences. | |
0.5 mL | |
Primary | |
Netrin-1 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction