Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R9B Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130228

Catalog No. PA130228


Add to cart

Description

Description

PA1-30228 detects Neurabin 2 from rat samples. PA1-30228 has been successfully used in immunoprecipitation, Western blot applications. The PA1-30228 immunogen is: GST fusion protein, corresponding to internal sequence amino acids 286-390 of Rat brain Neurabin 2.

interacts with protein phosphatase 1 and actin filaments; involved in regulation of synaptic transmission.
Specifications

Specifications

PPP1R9B
Polyclonal
Unconjugated
Ppp1r9b
Liquid
PPP1R9B
GST fusion protein, corresponding to internal sequence amino acids 286-390 of Rat brain Neurabin 2.
Protein G
RUO
Antibody
Polyclonal
Rat
Immunoprecipitation, Western Blot
1 mg/mL
0.1M tris glycine with 0.15M NaCl, 30% glycerol and 0.05% sodium azide; pH 7.4
Q6R891, O35274, Q96SB3
Neb2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
84687, 217124, 84686
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit