Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Neurexin 3/NRXN3, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128485

Catalog No. NBP128485


Add to cart

Description

Description

Neurexin 3/NRXN3 Polyclonal antibody specifically detects Neurexin 3/NRXN3 in Human, Mouse, Porcine, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

Neurexin 3/NRXN3
0.5 mg/mL
Peptide ELISA Detection limit 1:4000
NP_004787.2
C14orf60, chromosome 14 open reading frame 60, KIAA0743MGC176711, neurexin 3, neurexin III, Neurexin III-alpha, Neurexin III-beta, neurexin-3-beta
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize the reported isoform 1 (NP_004787.2).
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
NRXN3
Peptide with sequence C-PQERKDARSQKNT corresponding to internal region according to NP_004787.2.
Immunogen affinity purified
RUO
Primary
9369
Canine, Human, Mouse, Porcine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only