Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurexin I Mouse, Unlabeled, Clone: 17, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611882

Catalog No. BDB611882


Add to cart

Description

Description

Neurexins are a family of neuron-specific cell surface proteins that contain hundreds of isoforms. These isoforms are generated from two different promoters in three different genes that code for many splice-variant transcripts. In rat, neurexins include 160-200kDa α-neurexins and 90-100kDa β-neurexins, which include multipe isoforms identified as neurexin I, II, or III. Full-length α-neurexins contain three repeating domains that include a left arm (A) cell interaction module (LNS), a central EGF domain, and a right arm (B) LNS. β-neurexins are identical to α-neurexins in the C-terminal region, but lack the five LNS repeats and three EGF domains of α -neurexins. Neurexin Iα was first characterized as a Ca2+-dependent receptor for α-latrotoxin, the excitatory neurotoxin in black widow spider venom. The C-terminus of Neurexin Iα interacts with the synaptic vesicle protein, synaptotagmin, while the three repeated sequences in the extracellular region have homology to cell adhesion-related proteins. Thus, Neurexin Iα may be important for cell-cell interactions and synaptic protein localization during the neuronal differentiation.

Host Species: Mouse
Clone: 17
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Rat Neurein Iα aa. 1063-1184

Immunofluorescence, Western Blotting

Specifications

Specifications

Neurexin I
17
Unconjugated
Affinity Purified
Rat Neurein Iα aa. 1063-1184
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit