Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEUROD1, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human NEUROD1 protein.

Manufacturer:  Abnova Corporation H00004760D01

Catalog No. 89-002-906


Add to cart

Description

Description

This gene encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus. [provided by RefSeq

Sequence: MTKSYSESGLMGEPQPQGPPSWTDECLSSQDEEHEADKKEDDLEAMNAEEDSLRNGGEEEDEDEDLEEEEEEEEEDDDQKPKRRGPKKKKMTKARLERFKLRRMKANARERNRMHGLNAALDNLRKVVPCYSKTQKLSKIETLRLAKNYIWALSEILRSGKSPDLVSFVQTLCKGLSQPTTNLVAGCLQLNPRTFLPEQNQDMPPHLPTASASFPVHPYSYQSPGLPSPPYGTMDSSHVFHVKPPPHAYSAALEPFFESPLTDCTSPSFDGPLSPPLSINGNFSFKHEPSAEFEKNYAFTMHYPAATLAGAQSHGSIFSGTAAPRCEIPIDNIMSFDSHSHHERVMSAQLNAIFHD
Specifications

Specifications

NEUROD1
Unconjugated
No additive
BC009046
BETA2|BHF-1|NEUROD|bHLHa3
Rabbit
100 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunoprecipitation
Rabbit polyclonal antibody raised against a full-length human NEUROD1 protein.
NEUROD1
Liquid
NEUROD1
NEUROD1 (NP_002491.2, 1 a.a. ~ 356 a.a) full-length human protein.
RUO
Primary
4760
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit