Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF1 Monoclonal Antibody (McNFn27b), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA1085
Description
MA1-085 detects neurofibromin in human, rat, and mouse cells. MA1-085 has been successfully used in Western blot, and immunohistochemistry procedures. By Western blot, this antibody detects a ∽240-260 kDa protein. The MA1-085 immunogen is a synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin.
NF1 Monoclonal antibody specifically detects NF1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
NF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P21359, P97526, Q04690 | |
NF1 | |
Synthetic peptide corresponding to the N-terminal residues 27-41 of human neurofibromin. | |
200 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
McNFn27b | |
Unconjugated | |
NF1 | |
AW494271; DKFZp686J1293; E030030H24Rik; FLJ21220; neurofibromatosis 1; Neurofibromatosis type 1; neurofibromatosis-related protein NF-1; Neurofibromin; neurofibromin 1; Neurofibromin truncated; NF1; Nf-1; NFNS; VRNF; WSS | |
Mouse | |
Protein G | |
RUO | |
18015, 24592, 4763 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction