Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurofibromin Mouse anti-Human, Clone: NFn27b, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051430
Description
This Anti-Neurofibromin Antibody, clone NFn27b is validated for use in western blotting for the detection of Neurofibromin.Specifications
Neurofibromin | |
Monoclonal | |
Unconjugated | |
P21359 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. | |
IgG1 |
Western Blot | |
NFn27b | |
Purified immunoglobulin. Liquid in PBS. No preservative. | |
NF1, VRNF, DKFZp686J1293, NFNS, Neurofibromin, WSS | |
N-terminal peptide from human neurofibromin | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only