Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament Mouse anti-Mouse, Rat, Clone: 3G3, MilliporeSigma™

Mouse Monoclonal Antibody

Manufacturer:  MilliporeSigma 05848

Catalog No. 05-848-MI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This Anti-Neurofilament Antibody, heavy chain, KEP segment, clone 3G3 is validated for use in western blot, immunocytochemistry, immunohistochemistry for the detection of Neurofilament.
Specifications

Specifications

Neurofilament
3G3
Ascites in 0.05% sodium azide and 30% glycerol
Ascites
Murine
IgG
200μL
Primary
Murine, Rat
Immunocytochemistry, Immunohistochemistry, Western Blot
Unconjugated
NM_021076
NEFH, NF-H, KIAA0845, NFH
Trp-E fusion protein corresponding to residues 846-1072 of rat neurofilament heavy chain.
Ascites
−20°C for 24 months.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only