Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-H Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14133

Catalog No. PA14133


Add to cart

Description

Description

PA1-4133 detects heavy neurofilament subunit tail in human samples. PA1-4133 has been successfully used in Western blot procedures. The PA1-4133 immunogen is a synthetic peptide corresponding to residues S P E K A K S P E K A K S, which is a repeated motif of human NFH.

The human heavy neurofilament subunit (NFH) tail that contains a repeated motif, XKSPYK. Deletions in this repeat region are strongly implicated as a primary event in amyotrophic lateral sclerosis.
Specifications

Specifications

NF-H
Polyclonal
Unconjugated
NEFH
Liquid
NEFH
Synthetic peptide corresponding to residues S P E K A K S P E K A K S, which is a repeated motif of human NFH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P12036
NFH, NF-H, NHC, 200 kDa neurofilament protein, KIAA0845, Neurofilament heavy polypeptide, Neurofilament triplet H protein
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4744
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit