Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEUROG1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant NEUROG1.

Manufacturer:  Abnova Corporation H00004762A01

Catalog No. 89-003-042


Add to cart

Description

Description

Sequence: SEALLHSLRRSRRVKANDRERNRMHNLNAALDALRSVLPSFPDDTKLTKIETLRFAYNYIWALAETLRLADQGLPGGGARER
Specifications

Specifications

NEUROG1
Unconjugated
50% glycerol
NM_006161
AKA|Math4C|NEUROD3|bHLHa6|ngn1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA
Mouse polyclonal antibody raised against a partial recombinant NEUROG1.
NEUROG1
Liquid
NEUROG1
NEUROG1 (NP_006152, 82 a.a. ~ 163 a.a) partial recombinant protein with GST tag.
RUO
Primary
4762
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit