Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurogenin-3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP227115SS
Description
Neurogenin-3 Polyclonal specifically detects Neurogenin-3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Neurogenin-3 | |
Polyclonal | |
Unconjugated | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
NEUROG3 | |
A portion of amino acids 1-69 of mouse Neurog 3 was used as the immunogen. | |
0.025 mg | |
Stem Cells | |
50674 | |
Mouse | |
IgG |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
Immunohistochemistry-Paraffin 5ug/ml | |
Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen is 83% homologous in rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction