Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuroG3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511893

Catalog No. PIPA511893

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Neurogenin-3 belongs to a family of basic helix-loop-helix transcription factors involved in the determination of neural precursor cells in the neuroectoderm. Neurogenin-3 also required for the specification of a common precursor of the 4 pancreatic endocrine cell types. Defects in NEUROG3 are the cause of congenital malabsorptive diarrhea 4 (DIAR4). DIAR4 is an autosomal recessive disorder characterized by generalized malabsorption and a paucity of enteroendocrine cells.

Specifications

Specifications

Neurogenin3
Polyclonal
Unconjugated
NEUROG3
Purified
NEUROG3
KLH conjugated synthetic peptide between 40-69 amino acids from the N-terminal region of human Neurogenin3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
2mg/mL
PBS with 0.09% sodium azide
Q9Y4Z2, P70661
Neurogenin-3, Protein atonal homolog 5, NEUROG3, ATOH5, NGN3
Rabbit
IgG
400μL
Store at 2°-8°C for up to 6 months. For long term storage store at -20°C
Primary
50674
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit