Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLGN3 Mouse anti-Human, Clone: 566209, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524253

Catalog No. PIMA524253


Add to cart

Description

Description

100% cross-reactivity with recombit human (rh) Neuroligin 3 variant 2 and recombit rat Neuroligin 3 variant 2 and no cross-reactivity with rhNeuroligin 1 variant 2, rhNeuroligin 2, rhNeuroligin 4, recombit rat Neuroligin 1, 1 variant 2, 2 variant 2, or 3 variant 4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
Specifications

Specifications

NLGN3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9NZ94
neuroligin 3, KIAA1480, ASPGX1, AUTSX1, HNL3, NLGN3
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
54413
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
566209
Unconjugated
NLGN3
Lyophilized
NLGN3
Mouse myeloma cell line NS0-derived recombit human Neuroligin 3 Gln38-Leu709
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit