Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuromedin U-8 Rabbit anti-Chicken, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA136127

Catalog No. 01-672-285


Add to cart

Description

Description

Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. Once reconstituted, store at -20°C.

may induce uterine smooth muscle contraction and selective vasoconstriction.
Specifications

Specifications

Neuromedin U-8
Polyclonal
whole serum with 0.1% sodium azide
P34963, P12760
NMU
Synthetic porcine neuromedin U-8 conjugated to BSA.
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
422748, 63887
Immunohistochemistry (Frozen)
Unconjugated
Nmu
Lyophilized
Rabbit
IgG
RUO
Antibody
Polyclonal
Chicken, Rat
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit