Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSE, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121081

Catalog No. PIPA121081

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Enolase is a glycolytic enzyme catalyzing the reaction pathway between 2-phospho-glycerate and phosphoenol pyruvate. In mammals, enolase molecules are dimers composed of three distinct subunits (alpha, beta and gamma). The alpha subunit is expressed in most tissues and the beta subunit only in muscle. The gamma subunit is expressed primarily in neurons, in normal and in neoplastic neuroendocrine cells. Coexpression of NSE (neuron specific enolase) and chromogranin A is common in neuroendocrine neoplasms.

PA1-21081 detects NSE in Human samples.PA1-21081 has been successfully used in immunohistochemistry (FFPE) procedures. Formalin-fixed paraffin-embedded tissue sections require boiling in 10 mM citrate buffer (pH 6.0) prior to immunostaining.The PA1-21081 immunogen is a synthetic peptide corresponding to the C-terminal portion of human NSE.
Specifications

Specifications

Neuron Specific Enolase
Polyclonal
Unconjugated
ENO2
Purified
ENO2
Synthetic peptide corresponding to the C-terminal portion of human NSE.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
P09104
Neuron Specific Enolase, glycerate hydrolyase, Eno, ENOG, Neuronal Enolase, Neural Enolase
Rabbit
IgG
500μL
4° C, do not freeze
Primary
2026
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit