Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCS1 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1939

Catalog No. PA1939

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The neuronal calcium sensor (NCS) proteins are bind calcium through EF-hand motifs and are predominantly expressed in neurons. Photoreceptor cells express type A sensors and include the proteins recoverin, S-modulin, and visinin. Type B receptors are expressed in neurons and include NCS1 and the Drosophila homolog frequenin, and VILIP. NCS1 has been shown to regulate neurosecretion in a calcium-dependent manner, potentiates nitric oxide synthase activity, and can activate calcineurin and 3'-5' nucleotide phosphodiesterase.

PA1-939 detects neuronal calcium sensor-1 (NCS-1) from rat tissues.PA1-939 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~20kDa protein representing NCS-1 from rat brain extract.PA1-939 immunizing peptide corresponds to amino acid residues 161-190 from rat NCS-1. This sequence is completely conserved in mouse, rat, chicken and human NCS-1. There is a single amino acid substitution in this region for Xenopus NCS-1.
Specifications

Specifications

NCS1
Polyclonal
Unconjugated
Ncs1
Liquid
NCS1
Synthetic Peptide: D(161) G K L T L Q E F Q E G S K A D P S I V Q A L S L Y D G L V(190)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P62168
NCS1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
65153
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit