Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuronal Pentraxin 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176899
Description
Neuronal Pentraxin 2 Polyclonal specifically detects Neuronal Pentraxin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Neuronal Pentraxin 2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
NARP, neuronal activity-regulated pentaxin, neuronal pentraxin IIapexin, neuronal pentraxin-2, NP2, NP-II | |
Rabbit | |
47 kDa | |
0.1 mg | |
Stem Cell Markers | |
4885 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 0.5-4 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin | |
NP_002514 | |
NPTX2 | |
Antibody was raised against a 16 amino acid synthetic peptide near the center of the human NPTX2. The immunogen is located within amino acids 170 - 220 of NPTX2. Amino Acid Squence: LHNETSAHRQKTESTL | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction