Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuronal Pentraxin Mouse, Unlabeled, Clone: 22, BD

Mouse Monoclonal Antibody

$213.00 - $427.00

Specifications

Antigen Neuronal Pentraxin
Isotype IgG1
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610369
View Documents Promotion Details
BD Biosciences
610369
50μg
Each for $213.00
Add to cart
 
BDB610370
View Documents Promotion Details
BD Biosciences
610370
150μg
Each for $427.00
Add to cart
 
Description

Description

Neuronal pentraxin (NP) is a potential receptor for the snake venom toxin, taipoxin. The highest levels of NP mRNA are found in the cerebellum and the CA3 region of the hippocampus. More specifically, NP mRNA is expressed at high levels in Purkinje and granule neurons of the cerebellum and CA3 neurons of the hippocampus. Moderate levels are present in neurons of the hilus, dentate gyrus, CA1, cerebral cortex, medial habenula, anterior hypothalamic nuclei, olfactory cortex, and inferior olivary nuclei. NP mRNA is not expressed in glia of white or gray matter. It is thought that NP serves as a receptor for taipoxin and, after binding, NP-taipoxin is taken into presynaptic terminals. Alternatively, the binding of taipoxin by NP may represent a nonspecific mechanism to sequester toxins from synapses.

Specifications

Specifications

Neuronal Pentraxin
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
22
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit