Learn More
NPY Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136133
Catalog No. 01-672-289
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.
NPY is a peptide hormone; involved in regulating food intake
Specifications
Specifications
NPY
Polyclonal
whole serum with 0.1% sodium azide
P07808
Neuropeptide-Y
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Npy
Lyophilized
NPY
A synthetic peptide corresonding to residues 1-21 of NPY, conjugated to BSA
50 μL
4° C
Primary
24604