Learn More
NPY2R, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA141576
Description
NPY2 receptor is a member of GPCR1 family and is localized primarily on Neuropeptide Y-expressing neurons in the hypothalamic region of the brain. It binds specifically to the C-terminal of Neuropeptide Y and is responsible for regulation of presynaptic relase of noradrenaline. Reports suggest a probable role of NPY2 receptor in regulation of feeding behavior, energy homeostasis and also in regulating cardiovascular system. Variations in the NPY2 receptor genes have been linked to occurrence of Diabetes.
NPY2R is also expected to react with mouse and canine samples based on sequence homology.Suggested positive control: human kidney protein, kidney lysate.
Specifications
NPY2R | |
Polyclonal | |
Unconjugated | |
NPY2R | |
NPY2-R | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4887 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5mg/mL | |
1-3ug/mL | |
P49146 | |
NPY2R | |
Synthetic peptide corresponding to residues 350-400 of human NPY2-R. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.