Learn More
Kallikrein 6 Mouse anti-Human, Clone: 9A7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen LFMA0153
Catalog No. LFMA0153
Add to cart
Description
Description
A suggested positive control for this product is Bosc23 cells transfected with neurosin.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. The encoded enzyme is regulated by steroid hormones. In tissue culture, the enzyme has been found to generate amyloidogenic fragments from the amyloid precursor protein, suggesting a potential for involvement in Alzheimer′s disease. Multiple alternatively spliced transcript variants that encode different isoforms have been identified for this gene.
Specifications
Specifications
Kallikrein 6
Monoclonal
Unconjugated
KLK6
Liquid
KLK6
Recombit human protein purified from E.coli
Ammonium sulfate precipitation
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
9A7
HEPES with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide
Q92876
Bssp, Klk7, MGC9355, NEUROSIN, PRSS18, PRSS9, SP59, ZYME, hK6
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5653