Learn More
NTF3 Sheep anti-Chicken, Human, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA118388
Catalog No. 01-670-640
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18388 detects NT3 from human, rat, and chicken samples. PA1-18388 has been successfully used in immunohistochemistry, ELISA, Western blot, and inhibition assay procedures. The PA1-18388 immunogen is Recombinant human NT3. Reconstitute with 100 μL of distilled water.
The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintece of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
Specifications
Specifications
NTF3
Polyclonal
whole serum with no preservative
P25433, P20783, P18280
Neurotrophin-3, Neurotrophic factor, HDNF, Nerve growth factor 2, NGF-2, Ntf3, Ntf-3
Sheep
IgG
RUO
Antibody
Polyclonal
Chicken, Human, Rat
ELISA, Immunohistochemistry, Inhibition Assays, Western Blot
Unconjugated
NTF3
Lyophilized
NTF3
Recombit human NT3.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
428099, 4908, 81737