Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NTF3 Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA19520
Description
PA1-9520 detects NT3 in human and mouse samples. PA1-9520 has been successfully used in ELISA procedures. PA1-9520 immunogen is a synthetic peptide based on the human NT3 protein.
NTF3 Polyclonal antibody specifically detects NTF3 in Human, Mouse samples. It is validated for ELISA.Specifications
NTF3 | |
Polyclonal | |
Unconjugated | |
Ntf3 | |
AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3 | |
Chicken | |
IgY fraction | |
RUO | |
18205, 4908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P20181, P20783 | |
Ntf3 | |
Synthetic peptide based on the human NT3 protein. | |
500 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgY |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction