Learn More
MilliporeSigma™ anti-Neutrophil Elastase, Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ 4810011ML
Catalog No. 48-100-11ML
Add to cart
Description
Description
Specifically detects Neutrophil Elastase Clone: in Human samples, and it is validated for ELISA, Immunoelectrophoresis, Immunoprecipitation, Immuno Blotting
Specifications
Specifications
Neutrophil Elastase
Unconjugated
purified
human neutrophil elastase
1mL
−20°C
Polyclonal
Human
ELISA, Immuno Blotting, Immunoblot, Immunoelectrophoresis, Immunoprecipitation
In PBS, pH 7.5.
Rabbit
IgG
RUO
Primary
Recognizes the neutrophil elastase protein.