Learn More
Ly-6G/Ly-6C Monoclonal Antibody (NIMP-R14), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA140038
Description
This clone is highly specific for murine Ly-6G and Ly-6C. It has been successfully used to stain polymorphonuclear (PMN) cells and monocytes for fluorescent activated cell sorting and in frozen and paraffin sections. Treatment with this antibody in vivo leads to neutropenia and has inhibitory effect on local immune responses. Furthermore, it has been shown to be useful for depletion of neutrophils in mice. It depletes neutrophils as soon as 6 hours after injection and up to 6 days. The endotoxin concentration is < 24 EU/mg, determined with LAL Assay. Suggested positive control: Mouse neutrophils; Suggested negative control: Mouse thymocytes
Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Ly-6G/Ly-6C | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and no preservative | |
P0CW02, P0CW03, P35461 | |
Ly6c1, Ly6c2, Ly6g | |
Purified BALB/c mouse neutrophils. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
NIMP-R14 | |
Unconjugated | |
Ly6c1 | |
AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN | |
Rat | |
Protein G | |
RUO | |
100041546, 17067, 546644 | |
4° C | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.