Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Nexilin, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals 47120002
Description
Nexilin Polyclonal antibody specifically detects Nexilin in Human samples. It is validated for ELISA.Specifications
Nexilin | |
Unconjugated | |
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative | |
Affinity Purified | |
NEXN | |
This antibody is specific for the N Terminus Region of the target protein. | |
Immunogen affinity purified | |
RUO | |
Primary | |
91624 | |
Human |
ELISA | |
ELISA 1:100-1:2000 | |
Q0ZGT2 | |
CMD1CC, F-actin-binding protein, MGC104234, MGC138865, MGC138866, Nelin, NELINnexilin, nexilin (F actin binding protein) | |
Rabbit | |
IgG | |
0.05 mg | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
This product is specific for Human NEXN. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only