Learn More
NFAM1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523106
Catalog No. PIPA523106
Add to cart
Description
Description
The protein encoded by this gene is a type I membrane receptor that activates cytokine gene promoters such as the IL-13 and TNF-alpha promoters. The encoded protein contains an immunoreceptor tyrosine-based activation motif (ITAM) and is thought to regulate the signaling and development of B-cells.
Specifications
Specifications
NFAM1
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Q8NET5
CTA-126B4.4, CNAIP, bK126B4.4, NFAT activation molecule 1, NFAT-activating protein with ITAM motif 1, calcinerin/NFAT-activating ITAM-containing protein, calcineurin/NFAT-activating ITAM-containing protein
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Western Blot
Unconjugated
NFAM1
Liquid
NFAM1
Synthetic peptide (amino acids 250-265) of NFAM1 protein.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
150372