Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFAT-1 Mouse, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610703

 View more versions of this product

Catalog No. BDB610703


Add to cart

Description

Description

T cells are activated and induced to proliferate following binding of their respective antigen. The process of includes expression of genes that encode factors (i.e., cytokines) which regulate various cell types. Modulation of gene expression is conducted by an array of specific interactions between transcription factors and DNA. NFAT-1 (Nuclear Factor of Activated T cells) is a transcription factor that regulates expression of the interleukin-2 gene. Thus, NFAT-1 DNA binding activity is undetectable in resting cells, but increases during T-cell activation. NFAT-1, a protein of 921 amino acids, is part of an oligomeric transcription factor that also contains Fra-1 and JunB. NFAT-1 was initially described as a phosphoprotein and is dephosphorylated in activated T cells transformed with the leukemia virus HTLV-l.

Specifications

Specifications

NFAT-1
1
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
NF-ATc2; NFATP; KIAA0611
Affinity Purified
Aqueous buffered solution containing BSA
150μg
Cell Biology
Primary
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.