Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14195

Catalog No. PA14195


Add to cart

Description

Description

PA1-4195 detects NFATc1 in human samples. NFATc1 is also known as NFAT2 or NFATc. PA1-4195 has been successfully used in Western blot procedures. The PA1-4195 immunogen is a synthetic peptide corresponding to residues L(102) S S G H T R P D G A P A(114) of human NFATc1.

The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that coordinate the immune response. NFATc is the cytosolic component of NFAT and encoded by a separate gene. Expression of NFATc activates the interleukin (IL-2) promoter in non-T lymphocytes and is required for IL-2 gene expression.
Specifications

Specifications

NFATc1
Polyclonal
Unconjugated
NFATC1
Liquid
NFATC1
Peptide corresponding to residues L(102) S S G H T R P D G A P A(114) of human NFATc1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O95644
NFATC1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1, MGC138448, NFAT2, NFATc, NFATC, NF-ATc, NF-ATC, NF-ATc1, NFAT tra
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4772
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit