Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NFATC4, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136747

Catalog No. NBP136747


Add to cart

Description

Description

NFATC4 Polyclonal antibody specifically detects NFATC4 in Human, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

NFATC4
0.5 mg/mL
Peptide ELISA Detection limit 1:64000
NP_001129494.1
NF-AT3, NFAT3nuclear factor of activated T-cells, cytoplasmic 4, NFATc4, NF-ATc4, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4, T cell transcription factor NFAT3, T-cell transcription factor NFAT3
Goat
IgG
0.1 mg
Transcription Factors and Regulators, Wnt Signaling Pathway
Primary
4776
Canine, Human
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
NFATC4
Peptide with sequence C-EATVNRLQSNEVTLT corresponding to internal region according to NP_001129494.1, NP_004545.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both isoforms (NP_001129494.1; NP_004545.2).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only