Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528299
Description
Recommended positive controls: nuclear factor I/B tansfected 293T cell, mouse brain. Predicted reactivity: Mouse (99%), Rat (100%), Xenopus laevis (95%), Chicken (97%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NFIB Polyclonal antibody specifically detects NFIB in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
NFIB | |
Polyclonal | |
Unconjugated | |
NFIB | |
6720429L07Rik; CCAAT-box-binding transcription factor; CTF; E030026I10Rik; HMGIC/NFIB; NF1-B; NF-I/B; Nfib; NFI-B; NFIB2; NFIB3; NFI-RED; nuclear factor 1 B-type; nuclear factor 1/B; nuclear factor I B; nuclear factor I/B; olfactory epithelium nuclear factor I-B; TGGCA-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18028, 4781 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.57 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O00712, P97863 | |
NFIB | |
Recombinant fragment corresponding to a region within amino acids 36 and 318 of Human NFIB | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction