Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nuclear factor I/C (CCAAT-binding transcription factor), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human NFIC protein.

Manufacturer:  Abnova Corporation H00004782B01

Catalog No. 89-005-856


Add to cart

Description

Description

Sequence: MYSSPLCLTQDEFHPFIEALLPHVRAFAYTWFNLQARKRKYFKKHEKRMSKDEERAVKDELLGEKPEVKQKWASRLLAKLRKDIRPECREDFVLSITGKKAPGCVLSNPDQKGKMRRIDCLRQADKVWRLDLVMVILFKGIPLESTDGERLVKAAQCGHPVLCVQPHHIGVAVKELDLYLAYFVRERDAEQSGSPRTGMGSDQEDSKPITLDTTDFQESFVTSGVFSVTELIQVSRTPVVTGTGPNFSLGELQGHLAYDLNPASTGLRRTLPSTSSSGSKRHKSGSMEEDVDTSPGGDYYTSPSSPTSSSRNWTEDMEGGISSPVKKTEMDKSPFNSPSPQDSPRLSSFTQHHRPVIAVHSGIARSPHPSSALHFPTTSILPQTASTYFPHTAIRYPPHLNPQDPLKDLVSLACDPASQQPGPSWYLG
Specifications

Specifications

nuclear factor I/C (CCAAT-binding transcription factor)
Unconjugated
No additive
NM_005597.2
CTF|CTF5|MGC20153|NF-I|NFI
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human NFIC protein.
NFIC
Serum
NFIC
NFIC (NP_005588.2, 1 a.a. ~ 428 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
4782
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit