Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFIL3, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131434

Catalog No. PA131434

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Expression of interleukin-3 (IL3; MIM 147740) is restricted to activated T cells, natural killer (NK) cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site (Zhang et al., 1995 [PubMed 7565758]).

PA1-31434 detects NFIL3 from human samples.PA1-31434 has been successfully used in Western blot applications.The PA1-31434 immunogen is: Peptide with sequence RLIATQPISASDSG, from C-Terminus of the protein sequence according to NP_005375
Specifications

Specifications

NFIL3
Polyclonal
Unconjugated
NFIL3
Purified
NFIL3
Peptide with sequence RLIATQPISASDSG, from C-Terminus of the protein sequence according to NP_005375
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q16649, O08750, Q6IMZ0
E4BP4, IL3BP1, NF-IL3A, NFIL3A
Goat
IgG
50μg
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
4783
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit