Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFkB p65 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141089

Catalog No. 01-676-680


Add to cart

Description

Description

This antibody is predicted to react new world monkey, porcine, canine, avian, and gorilla based on sequence homology. Suggested positive control: antigen standard for NFKB1 (transient overexpression lysate), HeLa or RAW.

NF-kB (nuclear factor kB) regulates the expression of a large number of genes that play critical roles in apoptosis, viral replication, tumorigenesis, various autoimmune diseases and inflammation. The active nuclear form of the NF-kB transcription factor complex is composed of two DNA binding subunits, NF-kB p65 and p50, both of which share extensive N-terminal sequence homology with the v-rel oncogene product. N-terminal regions of p50 and p65 are critical for DNA binding and help determine the DNA-binding specificities of p50 and p65.
Specifications

Specifications

NFkB p65
Polyclonal
Unconjugated
RELA
Liquid
RELA
Synthetic peptide corresponding to residues 531-540 of human NFkB p65.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q04206, Q04207
p65
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19697, 309165, 5970
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit