Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

NFKBIL1 Rabbit anti-Amphibian, Bovine, Canine, Equine, Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523499

Catalog No. PIPA523499



Description

Description

Suggested positive control: HeLa.

NF-kappa-B inhibitor-like protein 1 (NFKBIL1), also named Ikb like and IkbL1, is a 381 amino acid member of the inhibitor of NF-kappaB (IkappaB) family of proteins. NFKBIL1 contains two ANK repeats and is expressed in the rheumatoid synovial lining and sub-lining layers and co-localized in CD68+ and CD3+, but not Factor VIII+ cells. It is reported that defects in NFKBIL1 expression causes of susceptibility to rheumatoid arthritis (RA), which is a systemic inflammatory disease with autoimmune features and affects the joints. Rheumatoid arthritis (RA) is characterized by inflammatory changes in the synovial membranes and articular structures, widespread fibrinoid degeneration of the collagen fibers in mesenchymal tissues, and by atrophy and rarefaction of bony structures.
Specifications

Specifications

NFKBIL1
Polyclonal
Unconjugated
NFKBIL1
Liquid
NFKBIL1
Synthetic peptide (331 RRYLRVQQVRWHPDRFLQR 349) of NFKBIL1 protein.
Protein A
RUO
Antibody
Polyclonal
Amphibian, Bovine, Canine, Equine, Human, Non-human primate
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9UBC1
DADB-70P7.2, IKBL, LST1, NFKBIL, I-kappa-B-like protein, NF-kappa-B inhibitor-like protein 1, ikappaBL, inhibitor of kappa B-like protein
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100050027, 4795, 481711, 514369
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit