Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFYA Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Catalog No. PIPA129725
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA129725 50 μg
1 options

Catalog No. PIPA129725

Supplier: Thermo Scientific PA129725

Rabbit Polyclonal Antibody

PA1-29738 detects Dog IgG H&L from canine samples. PA1-29738 has been successfully used in Western blot and ELISA applications. The PA1-29738 immunogen is Dog IgG from normal dog serum.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NFYA
Applications ELISA, Gel Shift, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene NFYA
Gene Accession No. P23511, P23708
Gene Symbols NFYA
Host Species Rabbit
Immunogen NF-Y (A subunit) peptide corresponding to a region near the N-terminus conjugated to KLH
Purification Method Ion-exchange chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18044, 4800
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.