Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGAL Monoclonal Antibody (10), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific ABS0431002
Description
ABS 043-10-02 binds free NGAL in mouse serum. This antibody will not cross react with human, pig, dog, or rat. ABS 043-10-02 may be used in Western blot and sandwich ELISA with ABS 043-29-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
NGAL Monoclonal antibody specifically detects NGAL in Mouse samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
NGAL | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P11672 | |
LCN2 | |
Recombinant mouse NGAL adsorbed onto aluminum hydroxide gel. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Western Blot | |
10 | |
Unconjugated | |
LCN2 | |
24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin | |
Rat | |
Protein A/G | |
RUO | |
16819 | |
4° C, store in dark | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction