Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGRN Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524132

Catalog No. PIPA524132


Add to cart

Description

Description

NGRN may be involved in neuronal differentiation.
Specifications

Specifications

NGRN
Polyclonal
PBS with 0.09% sodium azide
Q9NPE2
DSC92, mesenchymal stem cell protein DSC92, neugrin, neurite outgrowth associated protein, neurite outgrowth-associated protein, spinal cord-derived protein FI58G
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51335
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
NGRN
Liquid
NGRN
KLH conjugated synthetic peptide between 261-290 amino acids from the C-terminal region of human NGRN
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit