Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHP2L1, Polyclonal, Invitrogen™

Catalog No. PIPA128264
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA128264 50 μg
1 options

Catalog No. PIPA128264

Supplier: Thermo Scientific PA128264

Chicken Polyclonal Antibody

Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5' stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.

PA1-28264 detects NHP2L1 from human samples.PA1-28264 has been successfully used in Western blot applications.The PA1-28264 immunogen is: Synthetic peptide, corresponding to amino acids 1 - 128 of Human NHP2L1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHP2L1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene SNU13
Gene Accession No. P55769, P55770
Gene Alias CTA-216E10.8, 15.5K, FA-1, FA1, NHPX, OTK27, SNRNP15-5, SNU13, SPAG12, SSFA1
Gene Symbols Nhp2l1, SNU13
Host Species Chicken
Immunogen Synthetic peptide, corresponding to amino acids 1 - 128 of Human NHP2L1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4809
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.