Learn More
Nicalin Rabbit anti-Human, Mouse, Rat, Pig, Chimpanzee, Canine, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061030
Catalog No. 06-103-0MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-Nicalin Antibody is an antibody against Nicalin for use in western blotting and immunocytochemistry.
Specifications
Specifications
Nicalin
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Canine, Chimpanzee, Human, Murine, Porcine, Rat
Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
NCLN
KLH-conjugated linear peptide corresponding to human Nicalin.
100μg
Primary
NP_064555