Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Niemann-Pick C1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$180.00 - $421.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118]
Antigen Niemann-Pick C1
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB400148SS
View Documents
Novus Biologicals
NB400148SS
0.025 ml
Each for $180.00
Add to cart
 
NB400148
View Documents
Novus Biologicals
NB400148
0.1 ml
Each for $421.50
Add to cart
 
Description

Description

Niemann-Pick C1 Polyclonal antibody specifically detects Niemann-Pick C1 in Human, Mouse, Rat, Porcine, Chinese Hamster, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy, Knockout Validated.
Specifications

Specifications

Rabbit
A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118]
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
O15118
PBS with 0.1% Sodium Azide
IgG
Niemann-Pick C1
Affinity Purified
RUO
Human, mouse, rat and Chinese hamster and primate (PMID 22212234). Porcine reactivity reported in scientific literature (PMID: 21051527).
4864
1.0 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit