Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Niemann-Pick type C1 Like-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 23 publications
Supplier: Novus Biologicals NB400128SS
Description
Niemann-Pick type C1 Like-1 Polyclonal specifically detects Niemann-Pick type C1 Like-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Niemann-Pick type C1 Like-1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
NPC1L1 | |
A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 500-600). [Uniprot: Q6T3U3] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:50-1:100, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:100 | |
Niemann-Pick C1-like protein 1, NPC1 (Niemann-Pick disease, type C1, gene)-like 1, NPC11L1 | |
Rabbit | |
140 kDa | |
0.025 mL | |
Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
29881 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction