Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIF3L1, Mouse, MaxPab™ Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human NIF3L1 protein.

Manufacturer:  Abnova Corporation H00060491B01

Catalog No. 89-002-523


Add to cart

Description

Description

Sequence: MDLKALLSSLNDFASLSFAESWDNVGLLVEPSPPHTVNTLFLTNDLTEEVMEEVLQKKADLILSYHPPIFRPMKRITWNTWKERLVIRALENRVGIYSPHTAYDAAPQGVNNWLAKGLGACTSRPIHPSKAPNYPTEGNHRVEFNVNYTQDLDKVMSAVKGIDGVSVTSFSARTGNEEQTRINLNCTQKALMQVVDFLSRNKQLYQKTEILSLEKPLLLHTGMGRLCTLDESVSLATMIDRIKRHLKLSHIRLALGVGRTLESQVKVVALCAGSGSSVLQGVEADLYLTGEMSHHDTLDAASQGINVILCEHSNTERGFLSDLRDMLDSHLENKINIILSETDRDPLQVV
Specifications

Specifications

NIF3L1
Unconjugated
No additive
NM_021824.2
ALS2CR1|CALS-7|MDS015
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human NIF3L1 protein.
NIF3L1
Liquid
NIF3L1
NIF3L1 (NP_068596.2, 1 a.a. ~ 350 a.a) full-length human protein.
RUO
Primary
60491
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit