Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ninjurin 2 (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human NINJ2 protein.

Manufacturer:  Abnova Corporation H00004815B02

Catalog No. 89-005-865


Add to cart

Description

Description

Cell adhesion molecules mediate cell-to-cell and cell-to-extracellular matrix interactions during development, differentiation, and regeneration of the peripheral nervous system. NINJ2 is upregulated in Schwann cells in the distal nerve segment after peripheral nerve injury and can promote neurite outgrowth.[supplied by OMIM

Sequence: MAGLSRQLCALSHPKKAAETQTAEPGGAHAVCSRHPVRVKGLEGSEMESARENIDLQPGSSDPRSQPINLNHYATKKSVAESMLDVALFMSNAMRLKAVLEQGPSSHYYTTLVTLISLSLLLQVVIGVLLVVIARLNLNEVEKQWRLNQLNNAATILVFFTVVINVFITAFGAHKTGFLAARASRNPL
Specifications

Specifications

ninjurin 2
Unconjugated
No additive
NM_016533.4
FLJ56349
Mouse
50 uL
Cell Adhesion
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human NINJ2 protein.
NINJ2
Serum
NINJ2
NINJ2 (NP_057617.2, 1 a.a. ~ 188 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
4815
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit