Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninjurin Mouse, Unlabeled, Clone: 50, BD

Mouse Monoclonal Antibody

$427.00

Specifications

Antigen Ninjurin
Isotype IgG2a
Immunogen Human Ninjurin aa. 1-152
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610777
View Documents
BD Biosciences
610777
150μg
Each for $427.00
Add to cart
 
BDB610776
View Documents
BD Biosciences
610776
50μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Ninjurin is a protein whose expression is dramatically increased after sciatic nerve transection and crush injuries. The Ninjurin gene encodes for a protein of 152 amino acids with two putative transmembrane domains. Ninjurin was iodinated in vivo and promoted the aggregation of Jurkat cells expressing Ninjurin, indicating a portion of Ninjurin is exposed to the cell surface. The adhesive properties of Ninjurin were energy and temperature dependent, required Ca2+; and Mg2+, and the integrity of the cytoskeleton. Also, the adhesion domain of Ninjurin is located at the extracellular NH2-terminal domain. Although its predicted mass is 16kDa, Ninjurin migrates as a 18-22kDa protein in SDS-PAGE, depending on the cell line or tissue. mRNA analysis revealed that Ninjurin is widely expressed with the highest levels in liver, thymus, heart, and the lowest level in brain.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Ninjurin
Human Ninjurin aa. 1-152
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG2a
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
50
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit